Sino Biological™ CEACAM-5 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human CEACAM5 (NP_004354.2) (Met 1-Ala 685) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11077H08H5

CEACAM-5 RECOMBINANT HUMAN PROTEIN, HIS TAG

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15288175

Quantity Price
1 £ 55.0 / 5µg
EU Stock 3
Add to basket


Description and Specification

Specification

Gene Alias CD66e, CEA, DKFZp781M2392, CEACAM5
Expression System Mammalian
Format Lyophilized
Molecular Weight 72.8kDa
Purification Method PAGE
Purity ≥95% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Full Length/Recombinant
Protein Subtype Glycoproteins
Endotoxin Concentration <1 EU/μg
Common Name Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein
Gene Symbol CEACAM5
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 1048

Carcinoembryonic Antigen-Related Cell Adhesion Molecule 5 Protein (CEACAM5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.