missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CDH16 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CDH16 (NP_004053.1) (Met 1-Ala 786) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10915H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15297375

  • £209.25 / 25µg


EU Stock 3
Add to basket

Description

Description

Cadherin 16 KSP-Cadherin (CDH16), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of MCF-7 human breast adenocarcinoma cells; when 5 times 104 cells/well are added to Recombinant Human Cadherin 16 coated plates (0.8μg/mL with 100μL/well), approximately >50% will adhere after 1 hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
84.8kDa
5 x 5μg
Glycoproteins|Intercellular Adhesion Molecule
UNQ695/PRO1340
CDH16
Cell Structure/Organelle Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Caderin 16 KSP-Cadherin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.