missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CDH1 Recombinant Human Protein, His Tag

This recombinant protein is expressed from a DNA sequence encoding the extracellular domain of human CDH1 precusor (NP_004351.1) (Met 1-Ile 707) fused to the polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10204H08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15234905

  • £166.56 / 50µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Cadherin 1, type 1, E-cadherin (epithelial) (CDH1) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.&line-break;Background: Cadherins are calcium-dependent cell adhesion proteins which preferentially interact with themselves in a homophilic manner in connecting cells, and thus may contribute to the sorting of heterogeneous cell type. E-cadherin (E-Cad), also known as CDH1 and CD324, is the classical cadherin molecule and contains five cadherin domains of ∽110 aa each in the extracellular domain. E-Cad is expressed in the non-neural epithelial tissues as disulfide-linked homodimer, and plays a key role in the organization and integrity of most epithelial tissues, cell differentiation and tissue development. CDH1 interacts directly, via the cytoplasmic domain, with CTNNB1 or JUP to form the PSEN1/cadherin/catenin adhesion complex which connects to the actin skeleton through the actin binding of alpha-catenin.
  • N-terminal Sequence Analysis: Gln 23 and Asp 155
  • Activity: measured by the ability of the immobilized protein to support the adhesion of MCF-7 human breast adenocarcinoma cells; when cells are added to CDH1/ECAD coated plates (1.5μg/mL, 100μL/well), approximately 60-80 % will adhere specifically
  • Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
77kDa
50μg
Other Proteins
CDH1, Arc-1, CD324, CDHE, ECAD, LCAM, UVO
CDH1
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cadherin 1, type 1, E-cadherin (epithelial)
Assay Standard
Mammalian
Videos