missing translation for 'onlineSavingsMsg'
Learn More

CDC42 Recombinant Human Protein, GST Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Cys 188) of human CDC42 isoform 2 (P60953-2) fused to the GST tag at the N-terminus

Brand:  Invitrogen 10062H09E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465676

  • £56.02 / 5µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Cell division cycle 42 (CDC42), GTP binding protein of 25kDa, recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris buffer, 150mM NaCl, 0.5mM reduced Glutathione, pH 8.0; contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
998
>85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
GTPases
Cell division cycle 42
Assay Standard
E. coli
Lyophilized
48.1kDa
5μg
Recombinant
RP1-224A6.5, CDC42Hs, G25K
CDC42
Other Proteins
GST Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.