Learn More
CD96 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met 1-Met 503) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 11202H08H250
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Cluster of Differentiation 96 (CD96), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: 1. Measured by its ability to bind recombinant mouse PVR in a functional ELISA; 2. Measured by its ability to bind recombinant Human CD155 in a functional ELISA
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
55kDa | |
5 x 50μg | |
C-Terminal | |
<1 EU/μg | |
CD96 Molecule | |
His Tag | |
CD Proteins | |
≥85% by SDS-PAGE |
10225 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Truncated/Recombinant | |
CD96 | |
CD96 | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.