missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD96 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met 1-Met 503) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11202H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228365

  • £233.19 / 25µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Cluster of Differentiation 96 (CD96), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind recombinant mouse PVR in a functional ELISA; 2. Measured by its ability to bind recombinant Human CD155 in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10225
PAGE
5 x 5μg
C-Terminal
Other Proteins
CD96
CD96
CD Proteins
His Tag
Lyophilized
55kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
CD96 Molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.