missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD9 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the second extracellular domain (Ser 112-Ile 195) of human CD9 (NP_001760.1) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Brand:  Thermo Scientific™ 11029H08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15247595

  • £653.75 / 50µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

CD9 Molecule (CD9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD19 at 2μg/mL (100μL/well) can bind biotinylated human CD9 with a linear range of 0.032-0.8μg/mL; 2. Immobilized mouse CD19 at 20μg/mL (100μL/well) can bind biotinylated human CD9 with a linear range of 0.032-0.8μg/mL; 3. Immobilized human CD21 at 2μg/mL (100μL/well) can bind biotinylated human CD9, the EC50 of biotinylated human CD9 is 1.76μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
11kDa
50μg
Other Proteins
MIC3, TSPAN29, GIG2, CD9
CD9
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD9 Molecule
Assay Standard
Mammalian
Videos