missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD8A Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Tyr 196) of mouse CD8A (NP_001074579.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50389M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13426296

  • £327.92 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Cluster of Differentiation 8A (CD8A), recombinant ferret protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its ability to bind biotinylated recombinant human B2M in a functional ELISA; 2. Measured by its ability to bind biotinylated recombinant human FCGRT and B2M in a functional ELISA; 3. Measured by its ability to bind biotinylated recombinant human LCK in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12525
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD8A, BB154331, Ly-2, Ly-35, Ly-B, Lyt-2
CD8A
CD Proteins
His Tag
Lyophilized
20.3kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD84 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.