missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD86 (B7-2) Recombinant Mouse Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 245) of mouse CD86 precursor (NP_062261.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50068M03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486186

  • £187.38 / 50µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Cluster of Differentiation 86 (CD86/B7-2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD28 at 20μg/mL (100μL/well) can bind mouse CD86-Fc with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12524
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
CD86, B7, B7-2, B7.2, B70, CLS1, CD28l2, ETC-1, Ly-58, Ly58, MB7, MB7-2, TS/A-2
CD86
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
53.4kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD86 antigen
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.