missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD86 (B7-2) Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 239) of human CD86 (NP_008820.2) fused to the His-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10699H03H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455936

  • £884.85 / 250µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Cluster of Differentiation 86 (CD86/B7-2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD28-His at 1μg/well can bind human CD86-Fc with a linear range of 1.25-20.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
942
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
B7-2, B70, CD28LG2, LAB72, mgC34413
CD86
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
53.2kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD86 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.