missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD74 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Gln 73-Met 232) of human CD74 isoform b (NP_004346.1) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11091H07H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15298215

  • £140.54 / 25µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Cluster of Differentiation 74 (CD74), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD74 at 5μg/mL (100μL/well) can bind biotinylated human CTSL1 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
972
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
HLA Class II
DHLAG, HLADG, Ia-GAMMA
CD74
CD Proteins
His Tag
Lyophilized
20.4kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD74 molecule, major histocompatibility complex, class II
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.