missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD69 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD69 (NP_001772.1) extracellular domain (Ser 62-Lys 199) fused to a signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11150H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218315

  • £1013.94 / 250µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

CD69 Molecule (CD69), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
969
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
CD69 Molecule
Assay Standard
Mammalian
Lyophilized
17.4kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
CLEC2C, CD69
CD69
Other Proteins
His Tag
Videos