missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD5L Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD5L (NP_005885.1) (Met 1-Gly 347) fused to a C-terminal polyhistidine tag was expressed

Brand:  Thermo Scientific™ 10791H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227175

  • £127.01 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Cluster of Differentiation 5 Antigen-Like (CD5L), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
922
PAGE
5 x 5μg
C-Terminal
Other Proteins
UNQ203/PRO229, AIM, API6, PRO229, SP-ALPHA, Spalpha, CD5L
CD5L
CD Proteins
His Tag
Lyophilized
37.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
CD5 antigen-like
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.