missing translation for 'onlineSavingsMsg'
Learn More

CD5 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 371) of mouse CD5 (NP_031676.3) fused to polyhistidine tag at the C-terminus

Brand:  Invitrogen 50403M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13436296

  • £148.00 / 25µg

Estimated Shipment date
from Supplier 16-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cluster of Differentiation 5 (CD5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12507
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD5, Ly-1, Ly-12, Ly-A, Lyt-1
CD5
CD Proteins
His Tag
Lyophilized
39.4kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD5 antigen
Assay Standard
Mammalian
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.