missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD44 Recombinant Human Protein, His Tag

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 220) of human CD44 (P16070-1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 12211H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13486146

  • £749.52 / 250µg

EU Stock 2
Add to basket



Cluster of Differentiation 44 (CD44) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.&line-break;Background: CD44, short for Cluster of Differentiation 44, is an adhesion molecule of the hyaluronate receptor family. The adhesive function of CD44, like other molecules involved in inducible adhesion, is tightly regulated. Posttranslational modifications, isoform expression, aggregation state, and protein associations all can affect the ligand binding properties of CD44, and these can vary depending on the cell type and the activation state of the cell. The principal ligand of CD44 is hyaluronan, and CD44 is involved in the metabolism of this compound. Furthermore, an increasing quantity of evidence suggests that CD44 has various functions related to inflammatory disease. CD44 deficiency induces severe liver injury. CD44-hyaluronate mediates in lymphocyte T and monocyte adhesion to the endothelium, stimulates proinflammatory cytokine release from macrophages and participates in dedifferentiation phenotype of smooth muscle cells from contractile state to synthetic one. CD44 acts in atherosclerotic plaque destabilization and neointimal proliferation after percutaneous vascular interventions. CD44 participates in many stages of atherosclerosis. CD44 also is a cell membrane protein often overexpressed on tumor cells and, being both a cell-cell and cell-extracellular matrix adhesion protein, is well positioned to contribute to this process.
  • N-terminal Sequence Analysis: Gln 21
  • Activity: This recombinant protein has not been tested
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
AL133330.1, CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, mgC10468
CD Proteins
His Tag
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD44 molecule (Indian blood group)
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.