missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD40LG Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the soluble form of human CD40LG (NP_000065.1) (Met 113-Leu 261) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10239H01H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15214995

  • £274.83 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

CD40 ligand (CD40LG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD40 at 2μg/mL (100μL/well) can bind human CD40LG-Fc chimera with a linear range of 1.28-32ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
42.8kDa
50μg
TNF (Tumor Necrosis Factor)
RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
CD40LG
CD Proteins
IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD40 ligand
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.