missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD40LG (CD154) Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the soluble form of human CD40LG (NP_000065.1) (Glu 108-Leu 261) fused to an N-terminal Met

Brand:  Thermo Scientific™ 10239H08E25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15224995

  • £123.88 / 25µg

Call for latest availability
+44 (0)1509 555 500
Add to basket



CD40 Ligand (CD40LG), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD40 at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 15.6-500ng/mL; 2. Immobilized human CD40/Fc at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 7.8-125ng/mL
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
TNF (Tumor Necrosis Factor)|Other Proteins
CD40 ligand
Assay Standard
His Tag
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
RP3-527F8.3, CD154, CD40L, HIGM1, IGM, IMD3, T-BAM, TNFSF5, TRAP, gp39, hCD40L
E. coli

For Research Use Only. Not for use in diagnostic procedures.