missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD40 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 193) of human CD40 (NP_001241.1) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10774H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13475966

  • £911.92 / 250µg

Estimated Shipment
Add to basket



Cluster of Differentiation 40 (CD40), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD40 at 2μg/mL (100μL/well) can bind biotinylated human CD40LG with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
TNF Receptors
CD40, Bp50, CDW40, mgC9013, TNFRSF5, p50
CD Proteins
His Tag
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD40 molecule, TNF receptor superfamily member 5
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.