missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD4 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 390) of human CD4 (NP_000607.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10400H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13403019

  • £55.44 / 5µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Cluster of Differentiation 4 (CD4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its ability to bind to HIV-1 gp120 in a functional ELISA; 2. Measured by the ability of the immobilized protein to support the adhesion of HeLa human cervical epithelial carcinoma cells; when 5 x 104 cells/well are added to CD4 coated plates (2.5μg/mL and 100μL/well), approximately 60-80% will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
920
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD4mut
CD4
CD Proteins
His Tag
Lyophilized
42.2kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD4 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.