missing translation for 'onlineSavingsMsg'
Learn More

CD4 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 390) of human CD4 (NP_000607.1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10400H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13483009

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Cluster of Differentiation 4 (CD4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: 1. Measured by its ability to bind to HIV-1 gp120 in a functional ELISA; 2. Measured by the ability of the immobilized protein to support the adhesion of HeLa human cervical epithelial carcinoma cells; when 5 x 104 cells/well are added to CD4 coated plates (2.5μg/mL and 100μL/well), approximately 60-80% will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
42.2kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD4 molecule
His Tag
CD Proteins
Glycoproteins
920
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
CD4mut
CD4
Mammalian
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.