missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD4 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 390) of human CD4 (NP_000607.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10400H03H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13415876

  • £57.00 / 5µg


Estimated Shipment
10-05-2018
Add to basket

Description

Description

Cluster of Differentiation 4 (CD4) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in the study of protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.&line-break;Background: Human T-cell surface glycoprotein CD4, also known as T-cell surface antigen T4/Leu-3, is a single-pass type I membrane protein. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CD4 is a glycoprotein expressed on the surface of T helper cells, regulatory T cells, monocytes, macrophages, and dendritic cells. CD4 amplifies the signal generated by the TCR. CD4 interacts directly with MHC class II molecules on the surface of the antigen presenting cell via its extracellular domain. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.
  • N-terminal Sequence Analysis: Lys26
  • Activity: measured by its binding ability in a functional ELISA; immobilized human CD4-His at 2μg/well has been shown to bind biotinylated HIV-1 GP120 with a linear range of 0.078-1.25μg/mL
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
920
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD4mut
CD4
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
68.7kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD4 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.