missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD3E Recombinant Cynomolgus Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the Cynomolgus (Macaca fascicularis) CD3E extracellular domain (Met 1 – Asp 117) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 90047C02H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15339716

  • £1180.50 / 250µg


Estimated Shipment
10-07-2018
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

CD3 Molecule, epsilon (CD3E), recombinant cynomplgus protein is supplied as a lyophilized powder.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
CD3E
CD3E
CD Proteins
IgG1 Fc Tag
Lyophilized
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
CD3 Molecule, epsilon
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.