Learn More
CD38 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human CD38 (NP_001766.2) (Val 43-Ile 300) fused to a C-terminal polyhistidine tag
Brand: Invitrogen 10818H08H25
Code : NEW
Additional Details : Weight : 0.01000kg
Description
ADP-Ribosyl Cyclase (CD38), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
31.3kDa | |
5 x 5μg | |
C-Terminal | |
<1 EU/μg | |
CD38 Molecule | |
His Tag | |
CD Proteins | |
Other Proteins |
952 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Truncated/Recombinant | |
T10, CD38 | |
CD38 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.