missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD36 Recombinant Human Protein, His Tag

This recombinant protein is expressed from a DNA sequence encoding the extracellular domain (Gly 30-Asn 439) of human CD36 (NP_001001547.1) fused both to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

Brand:  Thermo Scientific™ 10752H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13405966

  • £56.47 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Cluster of Differentiation 36 (CD36) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure the study of protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.&line-break;Background: CD36, is integral membrane protein found on the surface of many cell types in vertebrate animals. It belongs to the CD36 family and is found on platelets, erythrocytes, monocytes, differentiated adipocytes, mammary epithelial cells, spleen cells and some skin microdermal endothelial cells. CD36 has also been implicated in hemostasis, thrombosis, malaria, inflammation, lipid metabolism and atherogenesis. CD36 seems to have numerous potential physiological functions. It binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. It directly mediates cytoadherence of Plasmodium falciparum parasitized erythrocytes and binds long chain fatty acids. CD36 may also function as a cell adhesion molecule and in the transport and/or as a regulator of fatty acid transport. Defects in CD36 are the cause of platelet glycoprotein IV deficiency also known as CD36 deficiency. As a receptor for thrombospondin 1, CD36 plays a role in the regulation of angiogenesis, which may be a therapeutic strategy for controlling the dissemination of malignant neoplasms.
  • N-terminal Sequence Analysis: Asp 31
  • Activity: measured by its binding ability in a functional ELISA; immobilized human CD36 at 20μg/mL (100μL/well) has been shown to bind biotinylated human RSPO1 with a linear range of 0.032-4μg/mL
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
948
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD36, CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3
CD36
CD Proteins
His Tag
Lyophilized
48kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD36 molecule (thrombospondin receptor)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.