Sino Biological™ CD300A Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD300A (Q9UGN4-1) extracellular domain (Met 1-Gln 178) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 12449H08H25

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15255925

Quantity Price
1 £ 122.0 / 25µg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Gene ID (Entrez) 11314
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol CD300A
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Protein Tag His Tag
Purity ≥85% by SDS-PAGE
Protein Subtype Other Proteins
Quantity 5 x 5μg
Termination C-Terminal
Structural Form Truncated/Recombinant
Protein Family CD Proteins
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
Common Name CD300A Molecule
Validated Application Assay Standard
Format Lyophilized
Molecular Weight 18.8kDa
Purification Method PAGE

Cluster of Differentiation 300A (CD300A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.