missing translation for 'onlineSavingsMsg'
Learn More

CD274 Recombinant Mouse Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 238) of mouse CD274 (NP_068693.1) fused to the C-terminal His-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 50010M03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15342526

  • £167.00 / 50µg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

CD274 Antigen (CD274), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse PD-1 at 10μg/mL (100μL/well) can bind mouse CD274 with a linear range of 3.2-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52.8kDa
50μg
Other Proteins
CD274, B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1
CD274
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD274 Antigen
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.