missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD244 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 221) of human CD244 (NP_057466.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10042H08H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15254535

  • £219.66 / 50µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Cluster of Differentiation 244 (CD244), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD244 at 2μg/mL (100μg/well) can bind human CD48, the EC50 of human CD48 is 0.39μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23.8kDa
50μg
Other Proteins
2B4, CD244, NAIL, nMrk, NKR2B4, SLAMF4
CD244
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD244 molecule, natural killer cell receptor 2B4
Assay Standard
Mammalian
Videos