missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD24 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human CD24 (NP_037362.1) without the pro peptide (Met 1-Gly 59) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11030H02H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15267595

  • £127.01 / 25µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Cluster of Differentiation 24 (CD24), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
100133941
PAGE
5 x 5μg
C-Terminal
Glycoproteins|Intercellular Adhesion Molecule
CD24A, FLJ22950, FLJ43543, mgC75043
CD24
CD Proteins
IgG1 Fc Tag
Lyophilized
30kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
CD 24 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.