missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD226 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD226 (NP_848802.2) (Met 1-Pro 254) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50232M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15332966

  • £224.86 / 50µg


Estimated Shipment
09-07-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

CD226 antigen (CD226), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse CD226 at 2μg/mL (100μL/well) can bind mouse PVR/CD155 with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28.2kDa
50μg
Glycoproteins|Intercellular Adhesion Molecule
BC051526, DNAM-1, DNAM1, Pta1, TLiSA1, Cd226
CD226
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD226 antigen
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.