missing translation for 'onlineSavingsMsg'
Learn More

CD22 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 687) of human CD22-β (P20273-1) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11958H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13466146

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Cluster of Differentiation 22 (CD22), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
76.6kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD22 molecule
His Tag
CD Proteins
Lectins & Leptins
933
50μg
Truncated/Recombinant/Cell Surface Protein
RP11-446H13.1, FLJ41758, FLJ43475, FLJ44957
CD22
Mammalian
Lyophilized
≥95% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.