missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD209 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminally truncated extracellular (Lys 62-Ala 404) domain of human CD209 (NP_066978.1) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10200H01H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13495796

  • £745.36 / 250µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Cluster of Differentiation 209 (CD209), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
30835
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Lectin Receptor
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, mgC129965, CD209
CD209
CD Proteins
IgG1 Fc Tag
Lyophilized
65.8kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD209 molecule
Assay Standard
Mammalian
Videos