missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD200R1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 (NP_067300.1) fused to a C-terminal polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50209M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13416266

  • £144.70 / 25µg

Estimated Shipment
Add to basket



Cluster of Differentiation 200 receptor 1 (CD200R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its ability to bind mouse CD200 in a functional ELISA; immobilized mouse CD200-His at 0.2μg/well can bind mouse CD200R1-His with a linear range of 0.8-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CD200R, Mox2r, OX2R
CD Proteins
His Tag
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD200 receptor 1
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.