missing translation for 'onlineSavingsMsg'
Learn More

CD200R1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 266) of human CD200R1 (AAI43394.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

£58.15 - £875.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD200R, HCRTR2, MOX2R, OX2R, CD200R1
Gene ID (Entrez) 131450
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13466116 View Documents Invitrogen
11218H03H5
5μg £58.15
5µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-04-2020
Please to purchase this item. Need a web account? Register with us today!
13436116 View Documents Invitrogen
11218H03H25
5 x 5μg £145.00
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-04-2020
Please to purchase this item. Need a web account? Register with us today!
13476116 View Documents Invitrogen
11218H03H50
50μg £247.00
50µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-04-2020
Please to purchase this item. Need a web account? Register with us today!
13456116 View Documents Invitrogen
11218H03H250
5 x 50μg £875.00
250µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-04-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cluster of Differentiation 200 Receptor 1 (CD200R1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD200 at 1μg/mL (100μL/well) can bind human CD200R1-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
131450
<1 EU/μg
CD200R, HCRTR2, MOX2R, OX2R, CD200R1

For Research Use Only. Not for use in diagnostic procedures.