missing translation for 'onlineSavingsMsg'
Learn More

CD200R1 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 266) of human CD200R1 (AAI43394.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 11218H03H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13466116

  • £58.15 / 5µg

Estimated Shipment
20-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cluster of Differentiation 200 Receptor 1 (CD200R1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human CD200 at 1μg/mL (100μL/well) can bind human CD200R1-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Glycoproteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
131450
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD200 receptor 1
His Tag/IgG1 Fc Tag
CD Proteins
Assay Standard
Lyophilized
54.9kDa
5μg
Truncated/Recombinant/Cell Surface Protein
CD200R, HCRTR2, MOX2R, OX2R, CD200R1
CD200R1
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.