missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD200 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of mouse CD200 (NP_001076017.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50074M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13456196

  • £57.26 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Cluster of Differentiation 200 (CD200), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD200 at 1μg/mL (100μL/well) can bind mouse CD200R1-Fc with a linear range of 1.5-200ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17470
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD200, Mox2, OX2
CD200
CD Proteins
His Tag
Lyophilized
24kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD200 antigen
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.