missing translation for 'onlineSavingsMsg'
Learn More

CD200 Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human CD200 (NP_005935.4) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10886H03H250

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13455996

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Cluster of Differentiation 200 (CD200), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD200R1 at 1μg/mL (100μL/well) can bind human CD200-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
50.5kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD200 molecule
His Tag/IgG1 Fc Tag
CD Proteins
Glycoproteins
4345
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
CD200, MOX1, MOX2, MRC, OX-2
CD200
Mammalian
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.