missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD200 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human CD200 (NP_005935.4) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10886H03H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13445996

  • £144.70 / 25µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Cluster of Differentiation 200 (CD200), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD200R1 at 1μg/mL (100μL/well) can bind human CD200-Fc with a linear range of 0.12-16ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4345
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD200, MOX1, MOX2, MRC, OX-2
CD200
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
50.5kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD200 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.