missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CD14 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of the extracellular domain (Thr 20-Cys 352) of human CD14 (NP_000582.1) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10073H01H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455686

  • £57.26 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Cluster of Differentiation 14 (CD14), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
929
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD14 antigen, monocyte differentiation antigen CD14
CD14
CD Proteins
IgG1 Fc Tag
Lyophilized
62.4kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD14 molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.