missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CCL21 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human CCL21 (O00585) (Met 1-Pro 134)

Brand:  Thermo Scientific™ 10477HNAB25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15226775

  • £169.04 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Chemokine (C-C Motif) Ligand 21 (CCL21), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CCL21 at 2μg/mL (100μL/well) can bind human IGFBP7 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in 40mM Tris, 0.3M NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12.3kDa
5 x 5μg
Other Cytokines
RP11-195F19.22-001, 6Ckine, CKb9, ECL, mgC34555, SCYA21, SLC, TCA4
CCL21
Other Proteins
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Chemokine (C-C motif) ligand 21
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.