missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CCL1 Recombinant Mouse Protein, N-hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the secreted form of mouse CCL1 isoform 1 (P10146-1) (Lys 24-Cys 92) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 50178M01H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15302876

  • £82.96 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Chemokine (C-C Motif) Ligand 1 (CCL1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20290
PAGE
5μg
N-Terminal
Other Cytokines
RP23-446K18.2, BF534335, I-309, Scya1, Tca-3
CCL1
Chemokines & Receptors
IgG1 Fc Tag
Lyophilized
36.2kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Chemokine (C-C motif) ligand 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.