missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CADM3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CADM3 (short isoform) (NP_001120645.1) extracellular domain (Met 1-His 330) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11214H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15208385

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance



Cell Adhesion Molecule 3 (CADM3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; human CADM3 immobilized at 0.8μg/mL, 100μL/well will mediate >30% C6 cell adhesion
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
Neuropeptides|Intercellular Adhesion Molecule
CADM3, CTA-134P22.1, BIgR, FLJ10698, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
Other Proteins
His Tag
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Cell adhesion molecule 3
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.