missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CA2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

Brand:  Thermo Scientific™ 10478H08E50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15206785

  • £183.22 / 50µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
30kDa
50μg
Other Proteins
Carbonic anhydrase II
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CA-II, CAII, Car2
CA2
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.