Sino Biological™ CA2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10478H08E5

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15286775

Quantity Price
1 £ 53.75 / 5µg
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Specification

Quantity 5μg
Protein Tag His Tag
Validated Application Assay Standard
Protein Subtype Other Proteins
Expression System E. coli
Molecular Weight 30kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Purity ≥90% by SDS-PAGE
Gene Alias CA-II, CAII, Car2
Protein Family Other Proteins
Common Name Carbonic anhydrase II
Gene Symbol CA2
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.