Sino Biological™ CA2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10478H08E25

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15266775

Quantity Price
1 £ 115.0 / 25µg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Quantity 5 x 5μg
Molecular Weight 30kDa
Expression System E. coli
Validated Application Assay Standard
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias CA-II, CAII, Car2
Protein Subtype Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Family Other Proteins
Protein Tag His Tag
Purity ≥90% by SDS-PAGE
Gene Symbol CA2
Common Name Carbonic anhydrase II
Format Lyophilized

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.