missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ C1QBP Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human C1QBP (NP_001203.1) (His 75-Gln 282) fused to two Met at N-terminus and a polyhistidinetag at the C-terminus

Brand:  Thermo Scientific™ 11874H08E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15285575

  • £123.88 / 25µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Complement component 1, Q subcomponent Binding protein (C1QB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
708
PAGE
5 x 5μg
C-Terminal
Complement
C1QBP, GC1QBP, HABP1, SF2p32, gC1Q-R, gC1qR, p32
C1QBP
Other Proteins
His Tag
Lyophilized
24.8kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Complement component 1, q subcomponent binding protein
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.