missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BTC Recombinant Mouse Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse BTC (NP_031594.1) (Met 1-Gln 118) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50340M03H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15383126

  • £68.72 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Betacellulin, epidermal growth factor family member (BTC), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells; the ED50 for this effect is typically 5-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50kDa
5μg
EGF (Epidermal Growth Factor)
Btc, Bcn
BTC
Other Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Betacellulin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.