missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BST1 (CD157) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human BST1 (NP_004325.2), Met 1-Lys 292, fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10060H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425676

  • £57.00 / 5µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

Bone marrow stromal cell antigen 1 (BST1/CD157), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
683
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD157, BST1
BST1
CD Proteins
His Tag
Lyophilized
31.4kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Bone marrow stromal cell antigen 1
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.