missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BSG (CD147) Recombinant Mouse Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 209) of mouse BSG isoform 2 (NP_001070652.1) fused to a C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50332M03H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13496276

  • £74.96 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12215
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
AI115436, AI325119, CD147, EMMPRIN, HT-7, Bsg
BSG
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
48.6kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Basigin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.