missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BSG (CD147) Recombinant Human Protein, DDDDK Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 205) of human BSG (NP_940991.1) fused to a DDDDK tag at the C-terminus

Brand:  Thermo Scientific™ 10186HCCH5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13485766

  • £57.00 / 5µg


Estimated Shipment
15-05-2018
Add to basket

Description

Description

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
682
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN
BSG
CD Proteins
DDDDK
Lyophilized
20.7kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Basigin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.