missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BMPR2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 151) of human BMPR2 (NP_001195.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10551H03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455906

  • £186.34 / 50µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Bone morphogenetic protein receptor type II (BMPR2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
659
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Serine/Threonine Kinases|Growth Factor Receptors
BMPR2, BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK
BMPR2
Growth Factors & Receptors
His Tag/IgG1 Fc Tag
Lyophilized
42kDa
50μg
Truncated/Recombinant
<1 EU/μg
Bone morphogenetic protein receptor, type II (serine/threonine kinase)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.